Learn More
Thermo Scientific™ Savant™ DNA SpeedVac™ Concentrators Rotors Available on GSA/VA Contract for Federal Government customers only.
Configure Thermo Scientific™ Savant™ DNA SpeedVac™ Concentrators Rotors with Savant DNA SpeedVac Concentrators.
Manufacturer:  Thermo Scientific™ RD24
 View more versions of this product
Catalog No. 20-548-103
  Check Availability
Add to cart
Specifications
Description
Configure Thermo Scientific™ Savant™ DNA SpeedVac™ Concentrators Rotors with Savant DNA SpeedVac Concentrators.
Easy to use circular rotors with different tube hole sizes to accommodate different sample sizes. These rotors require no additional accessories and fit directly into your concentrator.
Description & Specifications
Specifications
24 x 1.5mL microtubes
Traditional
DNA 120/1200P
24
1.5mL