Thermo Scientific™ Savant™ SPD111 SpeedVac™ Kits Available on GSA/VA Contract for Federal Government customers only.

Obtain the necessary components for molecular biology applications with Thermo Scientific™ Savant™ SPD111V P1 and P2 SpeedVac™ kits.

Manufacturer: Thermo Scientific™ SPD111VP2115

 View more versions of this product

Catalog No. 11-716-805

  Check Availability
Add to Cart

    Specifications

    Includes -104C Refrigerated Vapor Trap; High-Vacuum Pump; 40 x 1.5mL Rotor; Universal Tubing Package; Digital Vacuum Gauge; Chemical Trap Kit; Disposable Cartridge; Glass Condensation Flask; Vacuum Pump Lubricating oil; CryoCool heat transfer fluid (1L)
    Width (English) Exterior 32 in.
    Shipping Weight (English) 360 lb.
    Shipping Weight (Metric) 136kg
    Description SpeedVac Concentrator Kit, Basic Molecular Biology
    Electrical Requirements 115V 60Hz
    Height (English) Exterior 34 in.
    Height (Metric) Exterior 864mm
    Length (English) Exterior 82 in.
    Length (Metric) Exterior 2083mm
    Width (Metric) Exterior 813mm
    Dimensions (L x W x H) Exterior 82 x 32 x 34 in. (2083 x 813 x 864mm)
    Voltage 115V
    Hertz 60Hz

    Obtain the necessary components for molecular biology applications with Thermo Scientific™ Savant™ SPD111V P1 and P2 SpeedVac™ kits. Choose our modular SPD111V P1 for standard evaporation, or the SPD111V P2 when you want to lyophillize. All of our SpeedVac kits are complete systems, containing everything you need under one part number.

    Features:

    • Variable drying rate
    • Time/temperature display incorporating membrane touch pad switches
    • Safety coverlock
    • Manual/auto timed run
    • Standard, deep aluminum Teflon™ coated chamber
    • Standard glass safety lids

    Solvent and Solvent Combinations:

    SPD111V P1

    • Acetonitrile
    • Ethanol
    • Isopropanol
    • Methanol
    • Water
    • Acetonitrile and Water
    • Ethanol and Water
    • Methanol and Water

    SPD111V P2

    • Acetonitrile
    • Ethanol
    • Methanol
    • Water
    • Acetonitrile and Water
    • Ethanol and Water
    • Methanol and Water

    Includes:

    SPD111V P1 SpeedVac Kit

    • SpeedVac concentrator, basic molecular biology
    • Universal vacuum system with -50°C refrigerated vapor trap
    • Glass condensation flask
    • 40 x 1.5mL rotor
    • Tubing package including 12 ft. of thick-walled vacuum tubing, cutter, clamps and fittings, BV130 manual bleed valve and instructions
    • CryoCool heat transfer fluid (1L)

    SPD111V P2 SpeedVac Kit

    • SpeedVac concentrator, basic molecular biology
    • -104°C, ultra-low temperature refrigerated vapor trap
    • 40 x 1.5mL rotor
    • Universal tubing package
    • Digital vacuum gauge
    • Chemical trap kit
    • Disposable cartridge
    • Glass condensation flask
    • Vacuum pump lubricating oil
    • CryoCool heat transfer fluid (1L)

    Recommended for:

    • Drying biomolecules in HPLC fractions
    • Evaporting purification solvents
    • Drying extracts of natural products in drug assays
    • Lyophillization for all the above (SPD111V P2)

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit