Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Savant™ SPD111 SpeedVac™ Kits Available on GSA/VA Contract for Federal Government customers only.

Obtain the necessary components for molecular biology applications with Thermo Scientific? Savant? SPD111V P1 and P2 SpeedVac? kits.

Manufacturer:  Thermo Scientific™ SPD111VP1115

 View more versions of this product

Catalog No. 11-716-804


  Check Availability
Add to cart

Specifications

Description

Obtain the necessary components for molecular biology applications with Thermo Scientific™ Savant™ SPD111V P1 and P2 SpeedVac™ kits. Choose our modular SPD111V P1 for standard evaporation, or the SPD111V P2 when you want to lyophillize. All of our SpeedVac kits are complete systems, containing everything you need under one part number.

Features:

  • Variable drying rate
  • Time/temperature display incorporating membrane touch pad switches
  • Safety coverlock
  • Manual/auto timed run
  • Standard, deep aluminum Teflon™ coated chamber
  • Standard glass safety lids

Solvent and Solvent Combinations:

SPD111V P1

  • Acetonitrile
  • Ethanol
  • Isopropanol
  • Methanol
  • Water
  • Acetonitrile and Water
  • Ethanol and Water
  • Methanol and Water

SPD111V P2

  • Acetonitrile
  • Ethanol
  • Methanol
  • Water
  • Acetonitrile and Water
  • Ethanol and Water
  • Methanol and Water

Includes:

SPD111V P1 SpeedVac Kit

  • SpeedVac concentrator, basic molecular biology
  • Universal vacuum system with -50°C refrigerated vapor trap
  • Glass condensation flask
  • 40 x 1.5mL rotor
  • Tubing package including 12 ft. of thick-walled vacuum tubing, cutter, clamps and fittings, BV130 manual bleed valve and instructions
  • CryoCool heat transfer fluid (1L)

SPD111V P2 SpeedVac Kit

  • SpeedVac concentrator, basic molecular biology
  • -104°C, ultra-low temperature refrigerated vapor trap
  • 40 x 1.5mL rotor
  • Universal tubing package
  • Digital vacuum gauge
  • Chemical trap kit
  • Disposable cartridge
  • Glass condensation flask
  • Vacuum pump lubricating oil
  • CryoCool heat transfer fluid (1L)

Recommended for:

  • Drying biomolecules in HPLC fractions
  • Evaporting purification solvents
  • Drying extracts of natural products in drug assays
  • Lyophillization for all the above (SPD111V P2)
Description & Specifications

Specifications

Universal Vacuum System with Oil-Free Vacuum Pump and -50C Refrigerated Vapor Trap; Glass Condensation Flask; Tubing Package, CryoCool Heat Transfer Fluid (1L)
60Hz
864mm
2083mm
813mm
136kg
82 x 32 x 34 in. (2083 x 813 x 864mm)
115V
34 in.
82 in.
32 in.
360 lb.
SpeedVac Concentrator Kit, Basic Molecular Biology
115V 60Hz
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit