Learn More
Thermo Scientific™ Savant™ SpeedVac™ Rotors for Medium and Large Capacity Concentrators Available on GSA/VA Contract for Federal Government customers only.
Combine these rotors with medium and large capacity Thermo Scientific™ Savant™ SpeedVac™ Concentrators.
Manufacturer:  Thermo Scientific™ RH1220
 View more versions of this product
Catalog No. 20-548-123
  Check Availability
Add to cart
Specifications
Description
Combine Thermo Scientific™ Savant™ SpeedVac™ Rotors for Medium and Large Capacity Concentrators with Savant SpeedVac Concentrators.
Circular Rotors:
  • Easy to use
  • Multi-size tube holes accommodate different sample sizes
  • Require no additional accessories
  • Fit directly into your concentrator
Description & Specifications
Specifications
Vials: 12 (20 x 47, 20 x 60mm), Medium capacity models
RH12-20
20 x 47, 20 x 60mm
Vials
12
Traditional
ISS110, SPD1010, SPD111V, SPD121P, SPD131DDA