Learn More
Thermo Scientific™ Savant™ SpeedVac™ Rotors for Medium and Large Capacity Concentrator, 6 x 50mL conical centrifuge tubes, Medium capacity models

Combine these rotors with medium and large capacity Thermo Scientific™ Savant™ SpeedVac™ Concentrators.
Manufacturer: Thermo Scientific™ RH650
Description
Combine Thermo Scientific™ Savant™ SpeedVac™ Rotors for Medium and Large Capacity Concentrators with Savant SpeedVac Concentrators.
Circular Rotors:
- Easy to use
- Multi-size tube holes accommodate different sample sizes
- Require no additional accessories
- Fit directly into your concentrator
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Centrifuge Tubes: 6 x 50mL conical, Medium capacity models | |
SPD1030, SPD120, SPD130, SPD140DDA | |
RH6-50 | |
50 mL |
Rotor | |
6 50 mL Conical | |
Traditional |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.