Learn More
Thermo Scientific™ Savant™ SpeedVac™ SPD300 Large Capacity Vacuum Concentrator and Kits with Installation
Thermo Scientific™ SpeedVac™ Modular Vacuum Concentrators are designed to vaporate strong acids, bases and aggressive organic solvents
Manufacturer:  Thermo Scientific™ SPD300DDAA115INS
 View more versions of this product
Catalog No. 13-875-379INS
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Approved for combinatorial chemistry
  • Incorporate an intuitive programmable user display for enhanced usability.
Specifications
Specifications
UL Listed to US and Canadian requirements. CE marked
SpeedVac SPD300DDA Vacuum Concentrator Kit-No rotor
115 V 60 Hz