Learn More
Thermo Scientific™ Savant™ SpeedVac™ SPD300 Large Capacity Vacuum Concentrator and Kits with Installation
Thermo Scientific™ SpeedVac™ Modular Vacuum Concentrators are designed to vaporate strong acids, bases and aggressive organic solvents
$41600.00 - $42800.00
Specifications
Electrical Requirements 115 V 60 Hz
View More Specs
Products 3
Catalog Number Mfr. No. Description Certifications Price Quantity & Availability  
Catalog Number Mfr. No. Description Certifications Price Quantity & Availability  
13-875-375INS
View Documents
Thermo Scientific™
SPD300P1115INS
SpeedVac SPD300DDA Vacuum Concentrator Kit UL Listed to US and Canadian requirements. CE marked
Each for $42,800.00
Add to cart
13-875-377INS
View Documents
Thermo Scientific™
SPD300P2115INS
SpeedVac SPD300DDA Vacuum Concentrator Kit UL Listed to US and Canadian requirements. CE marked.This product is not available for sale in California. Please contact Customer Service for possible alternatives.
Each for $41,600.00
Add to cart
13-875-379INS
View Documents
Thermo Scientific™
SPD300DDAA115INS
SpeedVac SPD300DDA Vacuum Concentrator Kit-No rotor UL Listed to US and Canadian requirements. CE marked This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
  • Approved for combinatorial chemistry
  • Incorporate an intuitive programmable user display for enhanced usability.
Specifications
Specifications
115 V 60 Hz