Ansell™ Sawyer-Tower™ 66-670 Series Jackets
Full-coverage chemical protection with added safety of fire resistance
Manufacturer: Ansell™ 66670 M 105415
 View more versions of this product
Catalog No. 19-010-035
  Check Availability
Add to Cart
    Specifications
    Material CPC Nomex Trilaminate
    Arc Flash Rating 2
    Size Medium
    Length (English) 30 in.
    Resistance Chemicals and Fire
    Color Blue
    Product Type Chemical resistant jacket
  • Description & Specifications
Related Products
Product Content Feedback