Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SB 203186 hydrochloride, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg

Chemical Identifiers

207572-69-8
C16H20N2O2·HCl
1-Piperidinylethyl-1H-indole-3-carboxylate hydrochloride

5-HT4 antagonist

Potent 5-HT4 receptor antagonist with high affinity for human atrial 5-HT4 receptors.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 10mg
Formula Weight 308.81
Chemical Name or Material SB 203186 hydrochloride
Grade >99%

Recommended Storage : Store at Room Temperature