Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SB 203580, Tocris

Selective inhibitor of p38 MAPK

Manufacturer:  R&D Systems 1202/50

Quantity 50mg

 View more versions of this product

Catalog No. 12-025-0


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

>98%
377.44
SB 203580
C21H16FN3OS
4-[5-(4-Fluorophenyl)-2-[4-(methylsulfonyl)phenyl]-1H-imidazol-4-yl]pyridine

Specifications

50mg
>98%
377.44
SB 203580
152121-47-6
C21H16FN3OS
4-[5-(4-Fluorophenyl)-2-[4-(methylsulfonyl)phenyl]-1H-imidazol-4-yl]pyridine

Safety and Handling

Recommended Storage : Desiccate at +4°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit