Learn More
SB 205384, Tocris
GABAA receptor modulator; slows current decay
Manufacturer:  R&D Systems 1512/10
Quantity 10mg
 View more versions of this product
Catalog No. 15-121-0
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
>98%
330.4
SB 205384
C17H18N2O3S
4-Amino-5,6,7,8-tetrahydro-7-hydroxy-2-methyl-[1H]benzothieno[2,3-b]pyridine-3-carboxylic acid 2-butynyl ester
Specifications
160296-13-9
>98%
330.4
SB 205384
10mg
C17H18N2O3S
4-Amino-5,6,7,8-tetrahydro-7-hydroxy-2-methyl-[1H]benzothieno[2,3-b]pyridine-3-carboxylic acid 2-butynyl ester
Safety and Handling
Recommended Storage : Desiccate at Room Temperature