Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SB 205384, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
GABAA receptor modulator; slows current decay. Potentiates α6, α3 and α5-subunit-containing receptors (EC50 values are 280, 695 and 730 nM respectively). Displays little effect on receptors containing α1 or α2 subunits.
Specifications
Quantity | 50 mg |
Formula Weight | 330.4 |
Percent Purity | >98% |
Chemical Name or Material | SB 205384 |
Safety and Handling
Recommended Storage : Desiccate at Room Temperature
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title