SB 218795, Tocris

Potent, selective non-peptide NK3 antagonist

Manufacturer: R&D Systems 1376/50

Quantity 50mg

 View more versions of this product

Catalog No. 13-765-0

  Check Availability
Add to Cart

    Chemical Identifiers

    Assay Percent Range >99%
    Molecular Formula C25H20N2O3
    Formula Weight 396.44
    Synonym (R)-[[2-Phenyl-4-quinolinyl)carbonyl]amino]-methyl ester benzeneacetic acid
    Chemical Name or Material SB 218795

    Specifications

    Formula Weight 396.44
    Quantity 50mg
    Assay Percent Range >99%
    Molecular Formula C25H20N2O3
    CAS 174635-53-1
    Synonym (R)-[[2-Phenyl-4-quinolinyl)carbonyl]amino]-methyl ester benzeneacetic acid
    Recommended Storage Store at Room Temperature
    Chemical Name or Material SB 218795

    Store at Room Temperature

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit