Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SB 223412, Tocris Bioscience™

Catalog Number 467250
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
174636-32-9
C25H22N2O2
382.463
BIAVGWDGIJKWRM-FQEVSTJZSA-N
talnetant, talnetant inn, unii-cz3t9t146k, chembl10188, n-alpha-ethylbenzyl-3-hydroxy-2-phenylquinoline-4-carboxamide, 3-hydroxy-2-phenyl-n-1s-1-phenylpropyl-4-quinolinecarboxamide, 3-hydroxy-2-phenyl-n-1s-1-phenylpropyl quinoline-4-carboxamide, n-s-alpha-ethylbenzyl-3-hydroxy-2-phenylcinchoninamide, 4-quinolinecarboxamide, 3-hydroxy-2-phenyl-n-1s-1-phenylpropyl, d0k5hi
5311424
3-hydroxy-2-phenyl-N-[(1S)-1-phenylpropyl]quinoline-4-carboxamide
CCC(C1=CC=CC=C1)NC(=O)C2=C(C(=NC3=CC=CC=C32)C4=CC=CC=C4)O
This item is not returnable. View return policy
174636-32-9
C25H22N2O2
382.463
BIAVGWDGIJKWRM-FQEVSTJZSA-N
talnetant, talnetant inn, unii-cz3t9t146k, chembl10188, n-alpha-ethylbenzyl-3-hydroxy-2-phenylquinoline-4-carboxamide, 3-hydroxy-2-phenyl-n-1s-1-phenylpropyl-4-quinolinecarboxamide, 3-hydroxy-2-phenyl-n-1s-1-phenylpropyl quinoline-4-carboxamide, n-s-alpha-ethylbenzyl-3-hydroxy-2-phenylcinchoninamide, 4-quinolinecarboxamide, 3-hydroxy-2-phenyl-n-1s-1-phenylpropyl, d0k5hi
5311424
3-hydroxy-2-phenyl-N-[(1S)-1-phenylpropyl]quinoline-4-carboxamide
CCC(C1=CC=CC=C1)NC(=O)C2=C(C(=NC3=CC=CC=C32)C4=CC=CC=C4)O

Potent, selective non-peptide NK3 antagonist. Brain penetrant

Potent and selective non-peptide NK3 receptor antagonist (Ki values are 1, 144 and >100000 nM for human NK3, NK2 and NK1 receptors respectively). Selective over a panel of >60 receptors, enzymes and ion channels at concentrations of 1 or 10 μM. Inhibits NKB-induced Ca2+ mobilization in vitro (IC50 = 16.6 nM) and inhibits NK3-agonist-induced behavioral responses in vivo. Orally active and brain penetrant.
Quantity 50 mg
Formula Weight 382.45
Percent Purity >98%
Chemical Name or Material SB 223412
Recommended Storage Store at +4°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit