Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Alfa Aesar™ SB 225002
A potent, selective and competitive antagonist of the G-protein coupled receptor CXCR2
Manufacturer:  Alfa Aesar™ J65387LB0
 View more versions of this product
Catalog No. AAJ65387LB0
Order in Bulk
Request bulk or custom formats
Specifications
Specifications
5mg
−30° to −10°C
A potent, selective, and competitive antagonist of the G-protein coupled receptor CXCR2
C13H10BrN3O4
1-(2-Bromophenyl)-3-(2-hydroxy-4-nitrophenyl)urea
Soluble to 100mM in DMSO and to 50mM in ethanol
Yellow
Solid
182498-32-4
352.10g/mol
Moisture Sensitive
Signal Transduction Reagent