Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SB 243213 dihydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Selective 5-HT2C inverse agonist (pKb = 9.8). Displays selectivity over other 5-HT2 subtypes (pKi values are 6.8, 7.0 and 9.0 for 5-HT2A, 5-HT2B and 5-HT2C respectively); also exhibits >100-fold selectivity over 50 other receptors, ion channels and enzymes. Displays anxiolytic activity in rat models.
Specifications
Quantity | 10 mg |
Formula Weight | 501.33 |
Percent Purity | >99% |
Chemical Name or Material | SB 243213 dihydrochloride |
Safety and Handling
Recommended Storage | Store at +4°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction