Learn More
SB 268262, Tocris™
Selective non-peptide CGRP1 antagonist
Manufacturer:  R&D Systems 4314/10
Quantity 10mg
 View more versions of this product
Catalog No. 43-141-0
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
>99%
401.46
SB 268262
C18H15N3O4S2
N-Methyl-N-(2-methylphenyl)-3-nitro-4-(2-thiazolylsulfinyl)-benzamide
Specifications
217438-17-0
C18H15N3O4S2
N-Methyl-N-(2-methylphenyl)-3-nitro-4-(2-thiazolylsulfinyl)-benzamide
SB 268262
>99%
401.46
10mg
Safety and Handling
Recommended Storage : Store at Room Temperature