Tocris™ SB 328437

Potent and selective CCR3 antagonist

Manufacturer: Tocris™ 3650/50

 View more versions of this product

Catalog No. 36-505-0R

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only


    Specifications

    Inhibitors SB 328437
    Purity >99
    Molecular Weight 378.38
    Quantity 50mg
    Product Type SB 328437

    Potent and selective CCR3 antagonist (IC50 = 4nM). Displays > 2500-fold selectivity over C5aR, LTD4, CCR7, CXCR1 and CXCR2 receptors. Inhibits eotaxin-, eotaxin-2- and MCP-4-induced Ca2+ mobilization (IC50 values are 38, 35 and 20nM respectively) and inhibits eotaxin-, eotaxin-2- and MCP-4-induced eosinophil chemotaxis with similar potencies.

    • Chemical Name: N-(1-Naphthalenylcarbonyl)-4-nitro-L-phenylalanine methyl ester
    • Formula: C21H18N2O5
    • CAS Number: 247580-43-4
    • Solubility: Soluble to 100mM in DMSO
    • Purity: >99

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit