Tocris™ SB 657510

Selective urotensin-II (UT) receptor antagonist

Manufacturer: Tocris™ 3571/10

 View more versions of this product

Catalog No. 35-711-0R

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only


    Specifications

    Inhibitors SB 657510
    Purity >98
    Molecular Weight 505.81
    Quantity 10mg
    Product Type SB 657510

    Selective urotensin-II (UT) receptor antagonist (Ki values are 61, 17, 30, 65 and 56nM at human, monkey, cat, rat and mouse receptors respectively). Inhibits U-II-induced intracellular Ca2+ mobilization (IC50 = 180nM) and antagonizes the contractile action of U-II in isolated mammalian arteries and aortae (EC50 = 50 - 189nM).

    • Chemical Name: 2-Bromo-N-[4-chloro-3-[[(3R)-1-methyl-3-pyrrolidinyl]oxy]phenyl]-4,5-dimethoxybenzenesulfonamide
    • Formula: C19H22BrClN2O5S
    • CAS Number: 474960-44-6
    • Solubility: Soluble to 100mM in DMSO
    • Purity: >98

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit