Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SB 706375, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
High affinity, non-peptide antagonist of the urotensin-II (UT) receptor. Exhibits high affinity for mammalian UT receptors, including human, mouse and rat (Ki values are 9.3, 19.1 and 20.7 nM respectively, in HEK293 cells expressing recombinant UT receptors). Also inhibits binding of radiolabeled urotensin to endogenous human UT receptors (Ki = 5.4 nM in a whole-cell binding assay). Displays ≥100-fold selectivity for the human UT receptor over 86 different receptors, ion channels, enzymes, transporters and nuclear hormones.
Specifications
Quantity | 10 mg |
Formula Weight | 539.36 |
Percent Purity | >98% |
Chemical Name or Material | SB 706375 |
Safety and Handling
Recommended Storage | Store at Room Temperature |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction