Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris™ SB 743921 hydrochloride

Potent kinesin spindle protein (KSP) inhibitor

Manufacturer:  Tocris™ 5109/50

 View more versions of this product

Catalog No. 51-095-0


  Check Availability
Add to cart

Disclaimers:

For Research Use Only


Specifications

Description

Potent kinesin spindle protein (KSP) inhibitor (Ki = 0.1nM). Induces cell mitotic arrest and apoptosis in vitro. Inhibits the growth of a range of tumor cells in vitro, including colon (HCT 116), prostate (PC-3) and leukemia (K-562) cancer cell lines. Causes tumor regression in human tumor xenograft models in vivo, including colon (Colo205), lung (H69) and breast (MCF7) cancer cell xenografts.
  • Chemical Name: N-(3-Aminopropyl)-N-[(1R)-1-[7-chloro-4-oxo-3-(phenylmethyl)-4H-1-benzopyran-2-yl]-2-methylpropyl]-4-methylbenzamide hydrochloride
  • Formula: C31H33ClN2O3.HCl
  • CAS Number: 940929-33-9
  • Solubility: Soluble to 100mM in DMSO
  • Purity: >98
Description & Specifications

Specifications

SB 743921 hydrochloride
>98
50mg
SB 743921 hydrochloride
553.52
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit