Tocris™ SC 144 hydrochloride

gp130 inhibitor; blocks cytokine-triggered gp130 signaling

Manufacturer: Tocris™ 4963/50

 View more versions of this product

Catalog No. 49-635-0

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only


    Specifications

    Inhibitors SC 144 hydrochloride
    Purity >99
    Molecular Weight 358.76
    Quantity 50mg
    Product Type SC 144 hydrochloride

    Inhibitor of gp130 (IC50 = 0.72µM); blocks cytokine-triggered gp130 signaling. Induces gp130 phosphorylation and down regulates gp130 glycosylation; also abrogates STAT3 phosphorylation, nuclear translocation and inhibits the expression of downstream target genes. Delays tumor growth in a mouse xenograft model of human ovarian cancer. Orally active.

    • Chemical Name: 2-(7-Fluoropyrrolo[1,2-a]quinoxalin-4-yl) 2-pyrazinecarboxylic acid hydrazide hydrochloride
    • Formula: C16H11FN6O.HCl
    • CAS Number: 917497-70-2
    • Solubility: Soluble to 100mM in DMSO
    • Purity: >99

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit