Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SC 58125, Tocris Bioscience™

Catalog Number 289510
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
162054-19-5
C17H12F4N2O2S
384.349
JHBIMJKLBUMNAU-UHFFFAOYSA-N
115239
CHEBI:8983
5-(4-fluorophenyl)-1-(4-methylsulfonylphenyl)-3-(trifluoromethyl)pyrazole
CS(=O)(=O)C1=CC=C(C=C1)N2C(=CC(=N2)C(F)(F)F)C3=CC=C(C=C3)F
This item is not returnable. View return policy
162054-19-5
C17H12F4N2O2S
384.349
JHBIMJKLBUMNAU-UHFFFAOYSA-N
115239
CHEBI:8983
5-(4-fluorophenyl)-1-(4-methylsulfonylphenyl)-3-(trifluoromethyl)pyrazole
CS(=O)(=O)C1=CC=C(C=C1)N2C(=CC(=N2)C(F)(F)F)C3=CC=C(C=C3)F

Selective cyclooxygenase-2 (COX-2) inhibitor

Selective cyclooxygenase 2 (COX-2) inhibitor (IC50 values are 0.04 and >100 μM for COX-2 and COX-1 respectively). Anti-inflammatory; blocks edema and hyperalgesia in vivo following an inflammatory insult, without causing gastric mucosal damage. Also displays antitumor activity.
Quantity 10 mg
Synonym 1-4-methylsulfonyl phenyl-3-trifluoromethyl-5-4-fluorophenyl pyrazole, 5-4-fluorophenyl-1-4-methylsulfonylphenyl-3-trifluoromethyl pyrazole, 5-4-fluorophenyl-1-4-methylsulfonyl phenyl-3-trifluoromethyl-1h-pyrazole, 5-4-fluorophenyl-1-4-methylsulfonyl phenyl-3-trifluoromethyl pyrazole, 1h-pyrazole, 5-4-fluorophenyl-1-4-methylsulfonyl phenyl-3-trifluoromethyl, curator_000018, d07tzm, 1h-pyrazole, 5-4-fluorophenyl-1-04-methylsulfonyl phenyl-3-trifluoromethyl
Formula Weight 384.35
Percent Purity >98%
Chemical Name or Material SC 58125
Recommended Storage Store at +4°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit