Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SC 58125, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Selective cyclooxygenase 2 (COX-2) inhibitor (IC50 values are 0.04 and >100 μM for COX-2 and COX-1 respectively). Anti-inflammatory; blocks edema and hyperalgesia in vivo following an inflammatory insult, without causing gastric mucosal damage. Also displays antitumor activity.
Specifications
Quantity | 50 mg |
Synonym | 1-4-methylsulfonyl phenyl-3-trifluoromethyl-5-4-fluorophenyl pyrazole, 5-4-fluorophenyl-1-4-methylsulfonylphenyl-3-trifluoromethyl pyrazole, 5-4-fluorophenyl-1-4-methylsulfonyl phenyl-3-trifluoromethyl-1h-pyrazole, 5-4-fluorophenyl-1-4-methylsulfonyl phenyl-3-trifluoromethyl pyrazole, 1h-pyrazole, 5-4-fluorophenyl-1-4-methylsulfonyl phenyl-3-trifluoromethyl, curator_000018, d07tzm, 1h-pyrazole, 5-4-fluorophenyl-1-04-methylsulfonyl phenyl-3-trifluoromethyl |
Formula Weight | 384.35 |
Percent Purity | >98% |
Chemical Name or Material | SC 58125 |
Safety and Handling
Recommended Storage | Store at +4°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction