SC-9, Tocris

Protein kinase C activator

Manufacturer: R&D Systems 0433/10

Quantity 10mg

 View more versions of this product

Catalog No. 04-331-0

  Check Availability
Add to Cart

    Chemical Identifiers

    Assay Percent Range >99%
    Molecular Formula C22H24ClNO2S
    Formula Weight 401.95
    Synonym 5-Chloro-N-(6-phenylhexyl)-1-naphthalenesulfonamide
    Chemical Name or Material SC-9

    Specifications

    Quantity 10mg
    Formula Weight 401.95
    Assay Percent Range >99%
    Molecular Formula C22H24ClNO2S
    CAS 102649-78-5
    Synonym 5-Chloro-N-(6-phenylhexyl)-1-naphthalenesulfonamide
    Recommended Storage Store at Room Temperature
    Chemical Name or Material SC-9

    Store at Room Temperature

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit