Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SC35 Mouse anti-Amphibian, Drosophila, Human, Rat, Clone: SC-35, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 8 publications
Specifications
Antigen | SC35 |
---|---|
Clone | SC-35 |
Dilution | Western Blot 1:100-1:2000, ELISA Indirect, Immunocytochemistry/Immunofluorescence 1:2000, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin, Electron Microscopy 1:10-1:500, Knockout Validated |
Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunoassay |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB1001774SS
|
Novus Biologicals
NB1001774SS |
0.025 mL | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
NB1001774
|
Novus Biologicals
NB1001774 |
0.1 mL | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
SC35 Monoclonal specifically detects SC35 in Human, Mouse, Rat, Amphibian, Drosophila samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Electron Microscopy, Immunohistochemistry Free-Floating, Knockout Validated.Specifications
SC35 | |
Western Blot 1:100-1:2000, ELISA Indirect, Immunocytochemistry/Immunofluorescence 1:2000, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin, Electron Microscopy 1:10-1:500, Knockout Validated | |
Monoclonal | |
Mouse | |
Human, Mouse, Rat, Amphibian, Drosophila | |
PR264, SC35, SC-35, serine/arginine-rich splicing factor 2, SFRS2, SFRS2A, splicing component, 35 kDa, splicing factor SC35, splicing factor, arginine/serine-rich 2, SR splicing factor 2, SRp30b | |
SRSF2 | |
IgG1 | |
Unpurified | |
Recognizes a phospho-epitope on the non-snRNP (small nuclear ribonucleoprotein particles) factor SC-35. The antibody reacts with the splicing factor SC-35 and with the SC-35-related non-snRNP factor SF2/ASF. |
SC-35 | |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunoassay | |
Unconjugated | |
RUO | |
Ascites with 15mM Sodium Azide | |
6427 | |
Partially purified mammalian spliceosome. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title