Learn More
Brady™ Scaffolding Danger Tags: DO NOT USE THIS SCAFFOLD KEEP OFF THIS SCAFFOLD IS BEING ERECTED, TAKEN DOWN OR…
Identify facility equipment to your exact specifications. Brady™ Scaffolding Tags are durable two-sided tags with a write-on area that accepts most marking pens.
Manufacturer:  Brady™ 86738
Catalog No. 19-113-170
Add to cart
Description
Description
  • Heavy duty paper tags are made of a durable cardstock material
  • They provide an economical and convenient way to ID equipment or areas of your facility
  • Tag includes a write-on area that accepts most marking pens
  • Tags are appropriate for indoor use
Specifications
Specifications
Black on red
Scaffolding
Not laminated
Rectangle
Paper cardstock
Header: DANGER
4 in. x 7 in.
(100) 12 in cotton string ties
100/Pack
-40°F to 194°F
DO NOT USE THIS SCAFFOLD KEEP OFF THIS SCAFFOLD IS BEING ERECTED, TAKEN DOWN OR HAS BEEN FOUND DEFECTIVE DO NOT ALTER DATE: COMPETENT PERSON: SIGNATURE: COMMENTS: