Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCAI Mouse anti-Human, Clone: 7-A4, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP267920

Catalog No. NBP267920


Add to cart

Description

Description

SCAI Monoclonal antibody specifically detects SCAI in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

SCAI
7-A4
Unconjugated
TBS (pH7.4), 1%BSA, 40%Glycerol with 0.2% Sodium Azide
C9orf126, FLJ11758, FLJ36664, MGC120528, protein SCAI, suppressor of cancer cell invasion
Mouse
IgM
100μL
Primary
286205
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
2 mg/mL
Western Blot 1:1000-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200
Purified
SCAI
Full length protein
Protein A purified
Store at -20°C. Avoid freeze-thaw cycles.
Monoclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit