Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SCAMP2 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1203431UV
Description
SCAMP2 Polyclonal antibody specifically detects SCAMP2 in Human, Mouse, Hamster samples. It is validated for Western BlotSpecifications
SCAMP2 | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Hamster | |
Purified |
Western Blot | |
DyLight 350 | |
secretory carrier membrane protein 2secretory carrier-associated membrane protein 2 | |
Synthetic Peptide: M(1) S A F D T N P F A D P V D V N P(17) C | |
0.1 mL | |
Membrane Trafficking and Chaperones | |
10066 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction