Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCAMP2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548844

Catalog No. PA548844

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with mouse based on sequence homology.

This gene product belongs to the SCAMP family of proteins which are secretory carrier membrane proteins. They function as carriers to the cell surface in post-golgi recycling pathways. Different family members are highly related products of distinct genes, and are usually expressed together. These findings suggest that the SCAMPs may function at the same site during vesicular transport rather than in separate pathways.
Specifications

Specifications

SCAMP2
Polyclonal
PBS with 0.09% sodium azide
O15127
Secretory carrier-associated membrane protein 2, Secretory carrier membrane protein 2, SCAMP2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10066
Western Blot
Unconjugated
SCAMP2
Liquid
SCAMP2
KLH conjugated synthetic peptide between 119-147 amino acids from the Central region of human SCAMP2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit