Alfa Aesar™ Scandium(III) bromide, ultra dry, 99.99% (metals basis)

Manufacturer: Alfa Aesar™ 4719903

Quantity 1g

 View more versions of this product

Catalog No. AA4719903

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13465-59-3
    Formula Weight 284.67
    MDL Number MFCD00016324
    Chemical Name or Material Scandium(III) bromide, Ultra dry

    Specifications

    Formula Weight 284.67
    Packaging Ampule
    TSCA TSCA
    Physical Form Powder
    Sensitivity Hygroscopic
    MDL Number MFCD00016324
    CAS 13465-59-3
    Quantity 1g
    Storage Note 1
    • Ampouled under argon
    Chemical Name or Material Scandium(III) bromide, Ultra dry
    Assay 99.99%, (metals basis)

    • Ampouled under argon

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit