Alfa Aesar™ Scandium(III) chloride, anhydrous, 99.9% (REO)

Manufacturer: Alfa Aesar™ 1867706

Quantity 5g

 View more versions of this product

Catalog No. AA1867706

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10361-84-9
    Formula Weight 151.32
    MDL Number MFCD00011221
    Merck Index 14,8392
    Chemical Name or Material Scandium(III) chloride, Anhydrous

    Specifications

    Formula Weight 151.32
    TSCA TSCA
    Mesh Size −20 Mesh
    Sensitivity Hygroscopic
    Solubility Information Soluble in water; Insoluble in alcohol
    Odor Odorless
    MDL Number MFCD00011221
    Merck Index 14,8392
    Melting Point 939°C
    Quantity 5g
    Density 2.39g/mL
    CAS 10361-84-9
    Chemical Name or Material Scandium(III) chloride, Anhydrous
    Assay 99.9%, (REO)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit