Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Scandium(III) chloride, anhydrous, 99.9% (REO), Thermo Scientific Chemicals

Catalog Number p-7049688
Click to view available options
Quantity:
0.25 g
1 g
5 g
10361-84-9
MFCD00011221
10361-84-9
MFCD00011221

Scandium(III) chloride is a Prins cyclization catalyst. Used to form an intermetallic nickel-scandium compound film by potentiostatic cathodic reduction from molten salt mixtures.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 939°C
Quantity 0.25 g
Merck Index 14,8392
Solubility Information Soluble in water; Insoluble in alcohol
Formula Weight 151.32
Percent Purity 99.9%
Odor Odorless
Sensitivity Hygroscopic
Density 2.39 g/mL
Assay (REO)
Chemical Name or Material Scandium(III) chloride, Anhydrous
Show More Show Less
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 233-799-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit