Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Scandium(III) chloride hexahydrate, REacton™, 99.99% (REO)

Manufacturer:  Alfa Aesar™ 1121903

Quantity 1g

 View more versions of this product

Catalog No. AA1121903


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

20662-14-0
MFCD00149852
Scandium(III) chloride hexahydrate
259.41 (151.32 Anhydrous)
14,8392
Reacton™

Specifications

20662-14-0
259.41 (151.32 Anhydrous)
14,8392
1g
99.99%, (REO)
Crystalline Aggregates
MFCD00149852
Hygroscopic
Scandium(III) chloride hexahydrate
Reacton™

Safety and Handling

SDS & Safety Documents

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit