Alfa Aesar™ Scandium(III) nitrate hydrate, REacton™, 99.9% (REO)

Manufacturer: Alfa Aesar™ 6710403

Quantity 1g

 View more versions of this product

Catalog No. AA6710403

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13465-60-6
    Molecular Formula Sc(NO3)3·xH2O
    Formula Weight 230.97 (Anhydrous)
    MDL Number MFCD00011220
    UN Number UN1477
    Merck Index 14,8392
    Chemical Name or Material Scandium (III) nitrate hydrate
    Grade Reacton™

    Specifications

    Formula Weight 230.97 (Anhydrous)
    Health Hazard 3 GHS P Statement
    P221-P210-P220-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Keep/Store away from clothing/combustible materials.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Crystalline
    Molecular Formula Sc(NO3)3·xH2O
    CAS 13465-60-6
    UN Number UN1477
    Quantity 1g
    Health Hazard 2 GHS H Statement
    H272-H315-H319-H335
    May intensify fire
    oxidizer.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    TSCA TSCA
    Sensitivity Hygroscopic
    Merck Index 14,8392
    Color White
    Solubility Information Freely soluble in water and alcohol
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.
    MDL Number MFCD00011220
    Chemical Name or Material Scandium (III) nitrate hydrate
    Assay 99.9%, (REO)
    Grade Reacton™

    GHS H Statement
    H272-H315-H319-H335
    May intensify fire
    oxidizer.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P221-P210-P220-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Keep/Store away from clothing/combustible materials.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit