Scandium(III) oxide, 99.9% (REO), Alfa Aesar™, Reacton™

Manufacturer: Alfa Aesar 1121603

Quantity 1g

 View more versions of this product

Catalog No. AA1121603

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12060-08-1
    Physical Form Powder
    Assay Percent Range 99.9%
    Molecular Formula Sc2O3
    Formula Weight 137.91
    MDL Number MFCD00011223
    Chemical Name or Material Scandium(III) oxide

    Specifications

    Formula Weight 137.91
    Quantity 1g
    EINECS Number 235-042-0
    Physical Form Powder
    Odor Odorless
    Assay Percent Range 99.9%
    Molecular Formula Sc2O3
    MDL Number MFCD00011223
    CAS 12060-08-1
    Chemical Name or Material Scandium(III) oxide

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit