Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Scandium(III) trifluoromethanesulfonate, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
144026-79-9
C3F9O9S3Sc
492.17
MFCD00192433
HZXJVDYQRYYYOR-UHFFFAOYSA-K
scandium iii trifluoromethanesulfonate,scandium trifluoromethanesulfonate,scandium iii triflate,scandium triflate,sc otf 3,scandium 3+ ; trifluoromethanesulfonate,trifluoromethanesulfonic acid scandium iii salt,scandium iii trifluoromethanesulphonate,scandium tris trifluoromethanesulfonate,scandiumtriflate
2734571
scandium(3+);trifluoromethanesulfonate
C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Sc+3]
144026-79-9
C3F9O9S3Sc
492.17
MFCD00192433
HZXJVDYQRYYYOR-UHFFFAOYSA-K
scandium iii trifluoromethanesulfonate,scandium trifluoromethanesulfonate,scandium iii triflate,scandium triflate,sc otf 3,scandium 3+ ; trifluoromethanesulfonate,trifluoromethanesulfonic acid scandium iii salt,scandium iii trifluoromethanesulphonate,scandium tris trifluoromethanesulfonate,scandiumtriflate
2734571
scandium(3+);trifluoromethanesulfonate
C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Sc+3]
Melting Point 300°C
Color White
Quantity 1g
Packaging Glass bottle
Infrared Spectrum Authentic
Elemental Analysis 94% min.
Assay Percent Range 95%
Solubility Information Solubility in water: soluble
Formula Weight 492.17
Physical Form Powder
Percent Purity 95%
Chemical Name or Material Scandium(III) trifluoromethanesulfonate
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger