Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SCAP Antibody, Alexa Fluor™ 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP204113AF488
Description
SCAP Polyclonal specifically detects SCAP in Human samples. It is validated for Western Blot.Specifications
SCAP | |
Polyclonal | |
Western Blot | |
KIAA0199, PSEC0227, SREBF chaperone, SREBP cleavage-activating protein, sterol regulatory element binding protein cleavage-activating protein | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG |
Western Blot | |
Alexa Fluor 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SCAP | |
The immunogen recognized by this antibody maps to a region between residue 450 and 500 of human Sterol Regulatory Element Binding Protein Ceavage-Activating Protein using the numbering given in entry NP_036367.2 (GeneID 22937). | |
100 μL | |
Cardiovascular Biology, Cholesterol Metabolism, Diabetes Research, Lipid and Metabolism, Signal Transduction | |
22937 | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction