Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SCC112 Rabbit anti-Human, DyLight 594, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298627DL594
Description
SCC112 Polyclonal specifically detects SCC112 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
SCC112 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunofluorescence | |
DyLight 594 | |
Cell proliferation-inducing gene 54 protein, DKFZp686B19246, FLJ41012, KIAA0648MGC131948, PDS5, PDS5, regulator of cohesion maintenance, homolog A (S. cerevisiae), PIG54, SCC-112MGC161503, Sister chromatid cohesion protein 112, sister chromatid cohesion protein PDS5 homolog A | |
Produced in rabbits immunized with E. coli-derived Human SCC112 fragment. | |
0.1 mL | |
Cell Cycle and Replication, DNA Repair | |
23244 | |
Store at 4°C in the dark. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title