Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SCCPDH Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP160126
Description
SCCPDH Polyclonal specifically detects SCCPDH in Human samples. It is validated for Western Blot.Specifications
SCCPDH | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
Q8NBX0 | |
SCCPDH | |
Synthetic peptides corresponding to SCCPDH(saccharopine dehydrogenase (putative)) The peptide sequence was selected from the C terminal of SCCPDH. Peptide sequence FSFGYFSKQGPTQKQIDAASFTLTFFGQGYSQGTGTDKNKPNIKICTQVK. | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Human: 100%; Equine: 85%. | |
Human, Rat, Canine, Equine, Guinea Pig | |
IgG |
Western Blot | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
CGI-49, EC 1.5.1.9, FLJ43187, NET11, probable saccharopine dehydrogenase, RP11-439E19.2, saccharopine dehydrogenase (putative) | |
Rabbit | |
Affinity Purified | |
RUO | |
51097 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title