Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCD Polyclonal Antibody, Invitrogen™

Catalog No. PIPA124412
Change view
Click to view available options
Quantity:
100 μg
Catalog No. Quantity
PIPA124412 100 μg
1 options

Catalog No. PIPA124412

Supplier: Thermo Scientific PA124412

Rabbit Polyclonal Antibody

PA1-24412 detects acyl-CoA desaturase (SCD) in human samples. PA1-24412 has been successfully used in Western blot procedures. PA1-24412 immunogen is a synthetic peptide region of human SCD. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Stearoyl-CoA desaturase ( fatty acid desaturase, SCD) is expressed at high levels in several human tissues and is required for the biosynthesis of oleate (18:1) and palmitoleate (16:1). These monounsaturated fatty acids are the major components of phospholipids, triglycerides, wax esters, and cholesterol esters.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCD
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Gene SCD
Gene Accession No. O00767
Gene Alias PRO1933, FADS5, MSTP008, SCD1, SCDOS
Gene Symbols SCD
Host Species Rabbit
Immunogen Synthetic peptide corresponding to a region of human SCD.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6319
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.