Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCD Rabbit anti-Human, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Proteintech Group IncSupplier Diversity Partner 233931AP

Catalog No. 50-173-4066


Add to cart

Description

Description

Stearoyl-CoA desaturase., ,
Specifications

Specifications

SCD
0.52 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O00767
Acyl CoA desaturase, Delta(9) desaturase, FADS5, Fatty acid desaturase, MSTP008, SCD, SCD1, SCDOS, Stearoyl CoA desaturase
Rabbit
IgG
150 μL
-20°C
Primary
246074, 6319
Immunohistochemistry (Paraffin)
Unconjugated
SCD
Liquid
SCD
SCD Fusion Protein Ag13456
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Rat
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit