Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Scepter™ 3.0 Cell Counter 40 μm Sensors

The Scepter™ 3.0 sensor is engineered with a micro-fabricated, cell-sensing zone enabling discrimination by cell size and volume- down to sub-micron and sub-pL resolution. Coupled with precision liquid handling channels and electronics, the Scepter™ sensor accurately provides cell population statistics.

$205.00 - $1829.00

Specifications

Product Type Scepter™ 3.0 Cell Counter Sensors
Cell Size 5 to 15 μm
Operating Cycles 50,000 to 1,500,000 particles/mL
Particle Size 4 to 18 μm
For Use With (Application) Cell Counting
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PHCC340050
View Documents Promotion Details
MilliporeSigma™
PHCC340050
50 Pk.
Each for $205.00
Add to cart
 
PHCC340250
View Documents
MilliporeSigma™
PHCC340250
250 Pk.
Each for $965.00
Add to cart
 
PHCC340500
View Documents
MilliporeSigma™
PHCC340500
500 Pk.
Each for $1,829.00
Add to cart
 
Description

Description

  • For research use only
  • Provides fast, convenient method for counting cells and particles
  • Compatible with both the 40 and 60 μM sensors
  • Employs the Coulter Principle in a miniaturized, handheld, format enabling rapid cell counting, taking less than 30 seconds
  • Prepare a dilution of the cell culture of interest and use cell counter to aspirate a sample of this dilution into the sensor
  • Displays:
    • Cell concentration
    • Average cell size
    • Average cell volume
    • Histogram of size or volume distribution
Specifications

Specifications

Scepter™ 3.0 Cell Counter Sensors
50,000 to 1,500,000 particles/mL
Cell Counting
5 to 15 μm
4 to 18 μm
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit