Sceptor Industries SpinCon™ Advanced Air Sampler

Collects numerous chemical and biological agents

Manufacturer: Sceptor Industries SPNBTY

 View more versions of this product

Catalog No. 19-735-462

  Check Availability
Add to Cart

    Specifications

    For Use With (Equipment) SpinCon™ wet sampler
    Product Type Rechargeable battery
    Voltage 12V
    Includes Effective for collection of micron and sub-micron-sized particles such as molds, pollen, fungi, bacteria, viruses, and bacteriophages, as well as nonvolatile, semivolatile, and certain volatile chemical compounds

    Portable wet concentrator air sampler collects bioaerosols, airborne particulate matter, and soluble vapors, concentrating them into a sampling solution compatible with a wide range of chemical and biological analytical techniques. 
     
    Features
     
    • Samples air at a rate of 450L/min. (16cfm), producing a 10mL concentrated liquid sample
    • Patented wet concentrator design achieves concentration factors greater than 600%
    • Output stream can directly interface with a variety of analytical instruments, providing near–real-time results
    • Sample can also be examined using standard biological culture, particle/organism counting, microscopy, immunoassay, PCR assay1, and flow cytometry
    • Adjustable sampling times selectable from 5 minutes to 6 hours
    • Automatically detects presence of container and extracts sample into bottle
    • Uses phosphate-buffered saline solution in combination with sterile water, but accommodates other collection fluids
    • Offers integral control guided by LCD status display, allowing manual extraction and injection
    • Both interior fluid lines and exterior surfaces decontaminate with the use of dilute bleach solution
    • Operates within a temperature range from 2° to 50°C (36° to 122°F)

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit