Sceptor Industries SpinCon™ Advanced Air Sampler
Collects numerous chemical and biological agents
Manufacturer: Sceptor Industries SPNCASE
 View more versions of this product
Catalog No. 19-735-464
  Check Availability
Add to Cart
    Specifications
    Product Type Travel case
    For Use With (Application) Protection for sampler during transport
    For Use With (Equipment) SpinCon™ wet sampler
    Material Plastic
    Includes Effective for collection of micron and sub-micron-sized particles such as molds, pollen, fungi, bacteria, viruses, and bacteriophages, as well as nonvolatile, semivolatile, and certain volatile chemical compounds
    Portable wet concentrator air sampler collects bioaerosols, airborne particulate matter, and soluble vapors, concentrating them into a sampling solution compatible with a wide range of chemical and biological analytical techniques. 
    Features
    • Samples air at a rate of 450L/min. (16cfm), producing a 10mL concentrated liquid sample
    • Patented wet concentrator design achieves concentration factors greater than 600%
    • Output stream can directly interface with a variety of analytical instruments, providing near–real-time results
    • Sample can also be examined using standard biological culture, particle/organism counting, microscopy, immunoassay, PCR assay1, and flow cytometry
    • Adjustable sampling times selectable from 5 minutes to 6 hours
    • Automatically detects presence of container and extracts sample into bottle
    • Uses phosphate-buffered saline solution in combination with sterile water, but accommodates other collection fluids
    • Offers integral control guided by LCD status display, allowing manual extraction and injection
    • Both interior fluid lines and exterior surfaces decontaminate with the use of dilute bleach solution
    • Operates within a temperature range from 2° to 50°C (36° to 122°F)
  • Description & Specifications
Related Products
Product Content Feedback