Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SCF/c-kit Ligand Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $426.00
Specifications
Antigen | SCF/c-kit Ligand |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot 3-5ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP227162SS
![]() |
Novus Biologicals
NBP227162SS |
0.025 mg |
Each for $206.50
|
|
|||||
NBP227162
![]() |
Novus Biologicals
NBP227162 |
0.1 mg |
Each for $426.00
|
|
|||||
Description
SCF/c-kit Ligand Polyclonal specifically detects SCF/c-kit Ligand in Human samples. It is validated for Western Blot.Specifications
SCF/c-kit Ligand | |
Western Blot 3-5ug/ml | |
Polyclonal | |
Purified | |
RUO | |
Human | |
DKFZp686F2250, familial progressive hyperpigmentation 2, FPH2, KIT ligand, Kitl, KL-1, Mast cell growth factor, MGFSHEP7, SCFStem cell factor, SFc-Kit ligand, steel factor | |
KITLG | |
IgG | |
Protein A purified | |
The amino acid sequence used as immunogen is 100% homologous in mouse and 77% homologous in rat. |
0.5 mg/mL | |
Western Blot | |
Unconjugated | |
Rabbit | |
Biologically Active Proteins, Cancer, Embryonic Stem Cell Markers, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
4254 | |
A synthetic peptide corresponding to amino acids 121-158 of mouse kitl was used as immunogen, GenBank no gb AAA39378.1 . | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title