Learn More
SCF/c-kit Ligand Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody has been used in 8 publications
Supplier:  R&D Systems AF255SP
Catalog No. AF255SP
Add to Cart
This item is not returnable. View return policy
Description
Description
SCF/c-kit Ligand Polyclonal specifically detects SCF/c-kit Ligand in Human samples. It is validated for Western Blot, Neutralization.
Specifications
Specifications
SCF/c-kit Ligand
Polyclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
c-kit Ligand, DCUA, DFNA69, DKFZp686F2250, familial progressive hyperpigmentation 2, FPH2, FPHH, KIT ligand, Kitl, KITLG, KL-1, Mast cell growth factor, MGF, MGFSHEP7, SCFStem cell factor, SFc-Kit ligand, SHEP7, SLF, steel factor
Goat
Affinity Purified
RUO
4254
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Western Blot, Neutralization
LYOPH
Western Blot 0.1 ug/mL, Neutralization 0.05-0.3 ug/mL
P21583
KITLG
E. coli-derived recombinant human SCF/c-kit Ligand (R&D Systems, Catalog # 255-SC) Glu26-Ala189 Accession # P21583
25 μg
Primary
Detects human SCF/c-kit Ligand in direct ELISAs and Western blots. In direct ELISAs, less than 60% cross-reactivity with recombinant mouse SCF is observed.
Human
IgG