Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCF/c-kit Ligand Rabbit anti-Human, Alexa Fluor 350, Clone: 4, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP289504AF350

Catalog No. NB067133


Add to cart

Description

Description

SCF/c-kit Ligand Monoclonal antibody specifically detects SCF/c-kit Ligand in Human samples. It is validated for ELISA, ELISA
Specifications

Specifications

SCF/c-kit Ligand
4
ELISA, Sandwich ELISA Detection
Purified
Rabbit
IgG
0.1 mL
Biologically Active Proteins, Cancer, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Stem Cell Markers
Primary
4254
ELISA
Alexa Fluor 350
50mM Sodium Borate
DKFZp686F2250, familial progressive hyperpigmentation 2, FPH2, KIT ligand, Kitl, KL-1, Mast cell growth factor, MGFSHEP7, SCFStem cell factor, SFc-Kit ligand, steel factor
Recombinant Human SCF/c-kit Ligand protein
Protein A purified
RUO
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit