Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCFD1 Mouse anti-Canine, Human, Rat, Clone: OTI2E7, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA504662

Catalog No. 50-167-5304


Add to cart

Description

Description

SCFD1 is involved in vesicular transport between the endoplasmic reticulum and the Golgi.
Specifications

Specifications

SCFD1
Monoclonal
0.87 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q62991, Q8WVM8
C14orf163, RA410, SLY1, SLY1P, STXBP1L2
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
23256, 480281, 54350
Immunohistochemistry (Paraffin), Western Blot
OTI2E7
Unconjugated
SCFD1
Liquid
SCFD1
Full length human recombit protein of human SCFD1 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Canine, Human, Rat
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit