Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCGB1A1 Mouse anti-Human, Clone:3A8B8, Abnova™

Mouse monoclonal antibody raised against recombinant human SCGB1A1.

Manufacturer:  Abnova Corporation MAB17750

Catalog No. 89-334-605


Add to cart

Specifications

Specifications

SCGB1A1
3A8B8
secretoglobin, family 1A, member 1 (uteroglobin)
In PBS (0.05% sodium azide)
Liquid
Mouse
IgG1
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
ELISA, Flow Cytometry, Western Blot
Unconjugated
ELISA (1:10000)
Flow Cytometry (1:200-1:400)
Immunocytochemistry
Immunohistochemistry
Western Blot (1:500-1:2000)
The optimal working dilution should be determined by the end user.
SCGB1A1
CC10, CC16, CCSP, UGB
Recombinant protein corresponding to amino acids 26-91 of human SCGB1A1 from E. coli.
100μg
Antibody
7356
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit